1334006
USP
Hydroxyzine pamoate
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Hydroxyzine pamoate salt
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
hydroxyzine
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
ClC1=CC=C(C=C1)C(N2CCN(CCOCCO)CC2)C3=CC=CC=C3.O=C(O)C4=C(O)C(CC5=C(C=CC=C6)C6=CC(C(O)=O)=C5O)=C7C=CC=CC7=C4
InChI
1S/C23H16O6.C21H27ClN2O2/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;22-20-8-6-19(7-9-20)21(18-4-2-1-3-5-18)24-12-10-23(11-13-24)14-16-26-17-15-25/h1-10,24-25H,11H2,(H,26,27)(H,28,29);1-9,21,25H,10-17H2
InChI key
ASDOKGIIKXGMNB-UHFFFAOYSA-N
Gene Information
human ... HRH1(3269)
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Hydroxyzine Pamoate Capsules
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service