Skip to Content
Merck
All Photos(1)

Key Documents

1296803

USP

Gonadorelin hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Gonadorelin dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

6.53 MG
$713.00

$713.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
6.53 MG
$713.00

About This Item

Empirical Formula (Hill Notation):
C55H75N17O13 · 2HCl
CAS Number:
Molecular Weight:
1255.21
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$713.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

gonadorelin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[nH]1cnc(c1)C[C@H](NC(=O)[C@H]6NC(=O)CC6)C(=O)N[C@@H](Cc4c5c([nH]c4)cccc5)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccc(cc3)O)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCN=C(N)N)C(=O)N2[C@@H](CCC2)C(=O)NCC(=O)N

InChI

1S/C55H75N17O13/c1-29(2)19-38(49(80)67-37(9-5-17-60-55(57)58)54(85)72-18-6-10-43(72)53(84)62-25-44(56)75)66-46(77)26-63-47(78)39(20-30-11-13-33(74)14-12-30)68-52(83)42(27-73)71-50(81)40(21-31-23-61-35-8-4-3-7-34(31)35)69-51(82)41(22-32-24-59-28-64-32)70-48(79)36-15-16-45(76)65-36/h3-4,7-8,11-14,23-24,28-29,36-43,61,73-74H,5-6,9-10,15-22,25-27H2,1-2H3,(H2,56,75)(H,59,64)(H,62,84)(H,63,78)(H,65,76)(H,66,77)(H,67,80)(H,68,83)(H,69,82)(H,70,79)(H,71,81)(H4,57,58,60)/t36-,37-,38-,39-,40-,41-,42-,43-/m0/s1

InChI key

XLXSAKCOAKORKW-AQJXLSMYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Gonadorelin Hydrochloride is a synthetic polypeptide hygroscopic hormone that possesses the ability to trigger the secretion of luteinizing hormone from the hypothalamus. [1]

Application

Gonadorelin hydrochloride USP reference standard intended for use in specified quality tests and assays.

Also used to prepare standard, system suitability, and standard stock solution during the assay, and impurity analysis of Gonadorelin Hydrochloride and Gonadorelin for Injection. [1] [2]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gonadorelin for Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 2143-2143 (2018)
Gonadorelin Hydrochloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(4), 2146-2146 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service