Skip to Content
Merck
All Photos(1)

Key Documents

1294207

USP

Glucosamine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-(+)-Glucosamine hydrochloride, 2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$491.00

$491.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
200 MG
$491.00

About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
4157370
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$491.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

glucosamine

manufacturer/tradename

USP

mp

190-194 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4-,5-,6?;/m1./s1

InChI key

QKPLRMLTKYXDST-NSEZLWDYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glucosamine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Glucosamine and Chondroitin Sulfate Sodium Tablets
  • Glucosamine and Methylsulfonylmethane Tablets
  • Glucosamine Hydrochloride
  • Glucosamine Sulfate Potassium Chloride
  • Glucosamine Sulfate Sodium Chloride
  • Glucosamine Tablets
  • Glucosamine, Chondroitin Sulfate Sodium, and Methylsulfonylmethane Tablets

Biochem/physiol Actions

Glucosamine, an amino sugar, is the precursor of the hexosamine biosynthetic pathway leading to the formation of UDP-N-acetylglucosamine (UDP-GlcNAc), which is then used for making glycosaminoglycans, proteoglycans, and glycolipids. D-(+)-Glucosamine inhibits the coaggregation of Candida yeast species with the bacterial strain S. salivarius.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oshra Betzer et al.
ACS nano, 8(9), 9274-9285 (2014-08-19)
A critical problem in the development and implementation of stem cell-based therapy is the lack of reliable, noninvasive means to image and trace the cells post-transplantation and evaluate their biodistribution, final fate, and functionality. In this study, we developed a
Cuiling Zhong et al.
Nature communications, 11(1), 6330-6330 (2020-12-12)
Endothelial cell (EC) metabolism is thought to be one of the driving forces for angiogenesis. Here we report the identification of the hexosamine D-mannosamine (ManN) as an EC mitogen and survival factor for bovine and human microvascular EC, with an
Antonella Denise Losinno et al.
Autophagy, 17(2), 439-456 (2020-01-28)
Cruzipain, the major cysteine protease of the pathogenic protozoa Trypanosoma cruzi, is an important virulence factor that plays a key role in the parasite nutrition, differentiation and host cell infection. Cruzipain is synthesized as a zymogen, matured, and delivered to
Ji-Sun Hwang et al.
Metabolism: clinical and experimental, 64(3), 368-379 (2014-12-18)
This study investigated the potential of glucosamine (GlcN) to affect body weight gain and insulin sensitivity in mice normal and at risk for developing diabetes. Male C57BL/6J mice were fed either chow diet (CD) or a high fat diet (HFD)
Khadijeh Jamialahmadi et al.
Environmental toxicology and pharmacology, 38(1), 212-219 (2014-06-25)
Glucosamine (GlcN) is an important precursor in the biochemical synthesis of glycosylated proteins and lipids in human body. It gains importance because of its contribution to human health and its multiple biological and therapeutic effects. In this study, the in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service