Skip to Content
Merck
All Photos(1)

Key Documents

1285750

USP

Flurbiprofen

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-2-Fluoro-α-methyl-4-biphenylacetic acid, L-790,330

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$544.00

$544.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
200 MG
$544.00

About This Item

Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$544.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

flurbiprofen

manufacturer/tradename

USP

mp

110-112 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C(O)=O)c1ccc(c(F)c1)-c2ccccc2

InChI

1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)

InChI key

SYTBZMRGLBWNTM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flurbiprofen USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Flurbiprofen Tablets
  • Flurbiprofen Sodium
  • Flurbiprofen Sodium Ophthalmic Solution

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor; non-steroidal anti-inflammatory agent with antifungal activity. The S enantiomer inhibits prostaglandin synthesis and has both anti-inflammatory and analgesic activity, while the R enantiomer does not inhibit prostaglandin synthesis and displays only analgesic activity.
Fluibiprofen is a cyclooxygenase (COX) inhibitor, which is an enzyme responsible for the conversion of arachidonic acid to prostaglandin G2 (PGG2) and PGG2 to prostaglandin H2 (PGH2) in the prostaglandin synthesis pathway. This decreases the prostaglandins which cause inflammation, pain, swelling and fever. Flurbiprofen inhibits the activity of both COX-1 and -2. The S enantiomer inhibits prostaglandin synthesis and has both anti-inflammatory and analgesic activity, while the R enantiomer does not inhibit prostaglandin synthesis and displays only analgesic activity.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flurbiprofen Sodium Ophthalmic Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(3), 1982-1982 (2013)
Flurbiprofen Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(3), 1980-1980 (2015)
Flurbiprofen Sodium
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(3), 1981-1981 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service