Skip to Content
Merck
All Photos(1)

Key Documents

1254508

USP

Estriol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17β-triol, 16α-Hydroxyestradiol, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$544.00

$544.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$544.00

About This Item

Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
Beilstein:
2508172
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$544.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

estriol

manufacturer/tradename

USP

mp

280-282 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O

InChI

1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1

InChI key

PROQIPRRNZUXQM-ZXXIGWHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Estriol is found to be a major estrogen[1] in pregnant women.[2] It originates in the placenta from the estrogen precursors, made by the fetal adrenal gland and liver. Estriol levels are directly related to the delivery of the fetus and serve as an index of the fetal well-being.[2]

Application

Estriol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Estriol Compounded Vaginal Cream

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 17 $\beta$-estradiol in pharmaceutical preparation by UV spectrophotometry and high performance liquid chromatography methods
Yilmaz B and Kadioglu Y
Arabian Journal of Chemistry, 10(3), S1422-S1428 (2017)
"Handbook of Poylmer-Liquid Interaction Parameters and Solubility Parameters ", 27(22), 4431-4438 (2012)
Development and validation of a liquid chromatographic method for the simultaneous determination of estradiol, estriol, estrone, and progesterone in pharmaceutical preparations
Wilson P
Journal of AOAC (Association of Official Analytical Chemists) International, 92(3), 846-854 (2009)
Simultaneous determination of natural and synthetic estrogens by EKC using a novel microemulsion
Tripodi V, et al.
Electrophoresis, 27(22), 4431-4438 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service