Skip to Content
Merck

1237600

USP

Epoprostenol sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Prostaglandin I2 sodium salt, (5Z,9α,11α,13E,15S)-6,9-Epoxy-11,15-dihydroxyprosta-5,13-dien-1-oic acid sodium salt, Epoprostenol sodium salt, PGI2-Na, Prostacyclin sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

100 MG

$4,580.00

$4,580.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
C20H31NaO5
CAS Number:
Molecular Weight:
374.45
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
6472195

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C20H32O5.Na/c1-2-3-4-8-15(21)10-11-16-17(22)12-18-20(16)14(13-25-18)7-5-6-9-19(23)24;/h7,10-11,15-18,20-22H,2-6,8-9,12-13H2,1H3,(H,23,24);/q;+1/p-1/b11-10+,14-7-;/t15-,16-,17+,18-,20+;/m0./s1

SMILES string

CCCCC[C@H](O)/C=C/[C@H]1[C@H](O)C[C@@](O/2)([H])[C@]1([H])CC2=C\CCCC(O)=O.[Na]

InChI key

OURCVRVJQMLUNA-QFDVFERUSA-M

grade

pharmaceutical primary standard

API family

epoprostenol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
123761112131031102000
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

epoprostenol

API family

epoprostenol

API family

dinoprostone

API family

cephalothin

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Epoprostenol sodium USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Prostacyclin therapy improves hemodynamics in pulmonary arterial hypertension.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C P Wheeler-Jones et al.
The Biochemical journal, 315 ( Pt 1), 41-47 (1996-04-01)
The 14-3-3 proteins inhibit protein kinase C (PKC) activity in vitro and contain conserved sequences that resemble the pseudosubstrate domain of PKC and the C-terminus of the annexins. In the present study we have identified the isoforms of 14-3-3 in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service