1225601
USP
Doxercalciferol
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraene-1α,3β-diol, (5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraene-1,3-diol, 1α-Hydroxyergocalciferol, 1α-Hydroxyvitamin D2, 1α-OHD2, 1-α-Hydroxyvitamin D2, 1-Hydroxyergocalciferol, TSA 840
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
doxercalciferol
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C(\CCC[C@]12C)=C\C=C3\C[C@@H](O)C[C@H](O)C3=C
InChI
1S/C28H44O2/c1-18(2)19(3)9-10-20(4)25-13-14-26-22(8-7-15-28(25,26)6)11-12-23-16-24(29)17-27(30)21(23)5/h9-12,18-20,24-27,29-30H,5,7-8,13-17H2,1-4,6H3/b10-9+,22-11+,23-12-/t19-,20+,24+,25+,26-,27-,28+/m0/s1
InChI key
HKXBNHCUPKIYDM-CGMHZMFXSA-N
Gene Information
human ... VDR(7421)
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also used to prepare standard, and standard stock solutions for the assay, and impurity analysis in Doxercalciferol[1] by Liquid chromatography in conjunction with UV detector according to the United States Pharmacopeia monograph (USP).
Biochem/physiol Actions
Analysis Note
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service