1224664
USP
Docosyl ferulate
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(2E)-3-(4-Hydroxy-3-methoxyphenyl)- 2-propenoic acid docosyl ester, 1-Docosyl ferulate, Docosyl trans-ferulate
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
ferulic acid
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
O(C)c1c(ccc(c1)C=CC(=O)OCCCCCCCCCCCCCCCCCCCCCC)O
InChI
1S/C32H54O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-27-36-32(34)26-24-29-23-25-30(33)31(28-29)35-2/h23-26,28,33H,3-22,27H2,1-2H3
InChI key
USNYNNITUQSEEV-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also, for use with USP monograph such as:
- Pygeum Capsules
- Pygeum Extract
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service