Skip to Content
Merck
All Photos(1)

Documents

1193505

USP

Diethyl phthalate

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Beilstein:
1912500
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

7.66 (vs air)

vapor pressure

1 mmHg ( 100 °C)

API family

diethyl phthalate

autoignition temp.

854 °F

expl. lim.

0.75 %, 187 °F

manufacturer/tradename

USP

refractive index

n20/D 1.502 (lit.)

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCOC(=O)c1ccccc1C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3

InChI key

FLKPEMZONWLCSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diethyl phthalate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service