Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 200 mg | Check Cart for Availability | $576.00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
diclazuril
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Clc1ccc(cc1)C(C#N)c2c(Cl)cc(cc2Cl)N3N=CC(=O)NC3=O
InChI
1S/C17H9Cl3N4O2/c18-10-3-1-9(2-4-10)12(7-21)16-13(19)5-11(6-14(16)20)24-17(26)23-15(25)8-22-24/h1-6,8,12H,(H,23,25,26)
InChI key
ZSZFUDFOPOMEET-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade analytical standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename EDQM | manufacturer/tradename - | manufacturer/tradename EDQM |
| format neat | format - | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) forensics and toxicology | application(s) pharmaceutical (small molecule) |
| API family diclazuril | API family diclazuril | API family - | API family diclazuril |
Still not finding the right product?
Explore all of our products under Diclazuril
signalword
Warning
hcodes
Hazard Classifications
Repr. 2 - STOT RE 2
target_organs
Lungs,Liver,lymph node
Storage Class
11 - Combustible Solids
wgk
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1188560-200MG | 04061833843529 |

