Skip to Content
Merck
All Photos(1)

Key Documents

1180503

USP

Dextromethorphan

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO
CAS Number:
Molecular Weight:
271.40
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

dextromethorphan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N1([C@@H]2[C@@H]3[C@@](CC1)(CCCC3)c4c(ccc(c4)OC)C2)C

InChI

1S/C18H25NO/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18/h6-7,12,15,17H,3-5,8-11H2,1-2H3/t15-,17+,18+/m1/s1

InChI key

MKXZASYAUGDDCJ-NJAFHUGGSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextromethorphan USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Guenin et al.
Clinical drug investigation, 34(9), 609-616 (2014-07-17)
Dextromethorphan hydrobromide (DM) is a widely used antitussive. This study determined, for the first time, the basic pharmacokinetic profile of DM and its active metabolite, dextrorphan (DP) in children and adolescents. Thirty-eight male and female subjects at risk for developing
Rüdiger Kaspera et al.
Biochemical pharmacology, 91(1), 109-118 (2014-06-29)
Ritonavir, an HIV protease inhibitor, is successfully used for the prevention and treatment of HIV infections. Ritonavir pharmacokinetics are complicated by inhibition, induction and pharmacogenetics of cytochrome P450 (CYP) enzymes mediating its clearance. This investigation revealed that CYP2J2, along with
Jasbir D Upadhyaya et al.
PloS one, 9(10), e110373-e110373 (2014-10-24)
Activation of bitter taste receptors (T2Rs) in human airway smooth muscle cells leads to muscle relaxation and bronchodilation. This finding led to our hypothesis that T2Rs are expressed in human pulmonary artery smooth muscle cells and might be involved in
Sang Yoon Lee et al.
Toxicology letters, 229(1), 33-40 (2014-06-10)
Although cytochrome P450 inhibition is the major drug-drug interaction (DDI) mechanism in clinical pharmacotherapy, DDI of a number of well-established drugs have not been investigated. Rifampicin, isoniazid, pyrazinamide and ethambutol combination therapy inhibits clearance of theophylline in patients with tuberculosis.
Yuka Muroi et al.
Drug metabolism and pharmacokinetics, 29(5), 360-366 (2014-03-22)
Genetic variations in cytochrome P450 2D6 (CYP2D6) contribute to interindividual variability in the metabolism of clinically used drugs, e.g., tamoxifen. CYP2D6 is genetically polymorphic and is associated with large interindividual variations in therapeutic efficacy and drug toxicity. In this study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service