Skip to Content
Merck
All Photos(1)

Key Documents

1180004

USP

Dextroamphetamine sulfate

United States Pharmacopeia (USP) Reference Standard, monograph mol wt. 368.49 ((C9H13N)2 · H2SO4)

Synonym(s):

D-Amphetamine hemisulfate salt, (+)-α-Methylphenethylamine hemisulfate salt, Dextroamphetamine hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N · 0.5 H2SO4
CAS Number:
Molecular Weight:
184.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

dextroamphetamine

mol wt

monograph mol wt. 368.49 ((C9H13N)2 · H2SO4)

manufacturer/tradename

USP

drug control

USDEA Schedule II; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CC(N)Cc1ccccc1.CC(N)Cc2ccccc2

InChI

1S/2C9H13N.H2O4S/c2*1-8(10)7-9-5-3-2-4-6-9;1-5(2,3)4/h2*2-6,8H,7,10H2,1H3;(H2,1,2,3,4)/t2*8-;/m00./s1

InChI key

PYHRZPFZZDCOPH-QXGOIDDHSA-N

Gene Information

human ... SLC18A2(6571)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextroamphetamine sulfate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Amphetamine Sulfate
  • Amphetamine Sulfate Tablets
  • Dextroamphetamine Sulfate
  • Dextroamphetamine Sulfate Tablets
  • Phenylpropanolamine Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivia A Moody et al.
Frontiers in pharmacology, 11, 585356-585356 (2021-01-12)
In the United States, fentanyl causes approximately 60,000 drug overdose deaths each year. Fentanyl is also frequently administered as an analgesic in the perioperative setting, where respiratory depression remains a common clinical problem. Naloxone is an efficacious opioid antagonist, but

Protocols

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service