Skip to Content
Merck
All Photos(1)

Key Documents

1098209

USP

Cefuroxime sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cefuroxime sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$544.00

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$544.00

About This Item

Empirical Formula (Hill Notation):
C16H15N4NaO8S
CAS Number:
Molecular Weight:
446.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

cefuroxime

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(N)=O)=C(N2C1=O)C([O-])=O)c3ccco3

InChI

1S/C16H16N4O8S.Na/c1-26-19-9(8-3-2-4-27-8)12(21)18-10-13(22)20-11(15(23)24)7(5-28-16(17)25)6-29-14(10)20;/h2-4,10,14H,5-6H2,1H3,(H2,17,25)(H,18,21)(H,23,24);/q;+1/p-1/b19-9-;/t10-,14-;/m1./s1

InChI key

URDOHUPGIOGTKV-JTBFTWTJSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Liu et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(10), 831-836 (2014-09-18)
To compare the effects of topical nonsteroidal anti-inflammatory drugs on pupil dilation maintenance during phacoemulsification cataract surgery and quantify the relationships between pupil size change and nuclear hardness. This prospective randomized clinical observation study was single centered and double-masked. We
Zhenqiang Fang et al.
Journal of investigative surgery : the official journal of the Academy of Surgical Research, 27(6), 354-359 (2014-07-06)
Open nephroureterectomy (ONU) and bladder cuff resection (ONU-BCR) has been the gold standard of surgical treatment for upper urinary tract transitional cell carcinoma (UUT-TCC). The aim of this study is to introduce a modified total retroperitoneal laparoscopic nephroureterectomy (LNU) with
Lars Söderberg et al.
International journal of pharmaceutics, 484(1-2), 246-251 (2015-02-25)
Local irritation and inflammation at the site of administration are a common side effect following administration of parenteral formulations. Biological effects of surface (interfacial) activity in solutions are less well investigated than effects caused by other physico-chemical parameters such as
Astrit R Hamza et al.
World journal of gastroenterology, 20(40), 14841-14854 (2014-10-31)
To investigate the effect of meloxicam on the gut-liver axis after cirrhotic liver resection. Forty-four male Wistar rats were assigned to three groups: (1) control group (CG); (2) bile duct ligation with meloxicam treatment (BDL + M); and (3) bile
Mikkel Tøttrup et al.
Antimicrobial agents and chemotherapy, 58(6), 3200-3205 (2014-03-26)
Traditionally, the pharmacokinetics of antimicrobials in bone have been investigated using bone biopsy specimens, but this approach suffers from considerable methodological limitations. Consequently, new methods are needed. The objectives of this study were to assess the feasibility of microdialysis (MD)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service