Skip to Content
Merck
All Photos(1)

Key Documents

1080000

USP

Butacaine sulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Butacaine hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C18H30N2O2)2 · H2SO4
CAS Number:
Molecular Weight:
710.96
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

butacaine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/2C18H30N2O2.H2O4S/c2*1-3-5-12-20(13-6-4-2)14-7-15-22-18(21)16-8-10-17(19)11-9-16;1-5(2,3)4/h2*8-11H,3-7,12-15,19H2,1-2H3;(H2,1,2,3,4)

InChI key

VWZAGCZUPZKTET-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Butacaine sulfate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - Skin Corr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service