Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
200 mg | Check Cart for Availability | $565.00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
manufacturer/tradename
USP
bp
220 °C/4 mmHg (lit.)
mp
158-159 °C (lit.)
application(s)
pharmaceutical
format
neat
SMILES string
CC(C)(c1ccc(O)cc1)c2ccc(O)cc2
InChI
1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3
InChI key
IISBACLAFKSPIT-UHFFFAOYSA-N
General description
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename Manufactured by: Sigma-Aldrich Production GmbH, Switzerland | manufacturer/tradename - | manufacturer/tradename - |
| grade pharmaceutical primary standard | grade certified reference material, TraceCERT® | grade - | grade - |
| format neat | format neat | format - | format - |
| application(s) pharmaceutical | application(s) cleaning products | application(s) - | application(s) - |
| bp 220 °C/4 mmHg (lit.) | bp 220 °C/4 mmHg (lit.) | bp 220 °C/4 mmHg (lit.) | bp 220 °C/4 mmHg (lit.) |
| mp 158-159 °C (lit.) | mp 158-159 °C (lit.) | mp 158-159 °C (lit.) | mp 158-159 °C (lit.) |
Still not finding the right product?
Explore all of our products under Bisphenol A
signalword
Danger
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - ED ENV 1 - ED HH 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




