Skip to Content
Merck
All Photos(1)

Key Documents

1075203

USP

Bis(2-ethylhexyl) maleate

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$1,730.00

$1,730.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
$1,730.00

About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2O2CCH=CHCO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
340.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,730.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

bis(2-ethylhexyl) maleate

manufacturer/tradename

USP

refractive index

n20/D 1.455 (lit.)

density

0.944 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(CC)COC(=O)\C=C/C(=O)OCC(CC)CCCC

InChI

1S/C20H36O4/c1-5-9-11-17(7-3)15-23-19(21)13-14-20(22)24-16-18(8-4)12-10-6-2/h13-14,17-18H,5-12,15-16H2,1-4H3/b14-13-

InChI key

ROPXFXOUUANXRR-YPKPFQOOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-ethylhexyl) maleate is a maleate monomer used in polymerization.[1]

Application

Bis(2-ethylhexyl) maleate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymers from 1, 2-disubstituted ethylenic monomers, 3. Homopolymers from alkyl maleates: further results of monomer-isomerization radical polymerization.
Otsu, Takayuki, Osamu Ito, and Naoyuki Toyoda.
Makromol. Chem., Rapid Commun., 2.12, 729-732 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service