1048663
USP
Benazepril Related Compound E
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(3S)-3-Amino-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid monohydrochloride
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
benazepril
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
N1(c2c(cccc2)CC[C@@H](C1=O)N)CC(=O)O
InChI
1S/C12H14N2O3/c13-9-6-5-8-3-1-2-4-10(8)14(12(9)17)7-11(15)16/h1-4,9H,5-7,13H2,(H,15,16)/t9-/m0/s1
InChI key
JTCTZPXAQOMDEI-VIFPVBQESA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service