Skip to Content
Merck

Skip To

1034512

USP

α-Amylcinnamyl alcohol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Benzylidene-1-heptanol

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
C6H5CH=C[(CH2)4CH3]CH2OH
CAS Number:
Molecular Weight:
204.31
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3127316


grade

pharmaceutical primary standard

agency

USP

API family

alpha-amylcinnamyl alcohol

manufacturer/tradename

USP

refractive index

n20/D 1.519 (lit.)

bp

141-143 °C/5 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC\C(CO)=C/c1ccccc1

InChI

1S/C14H20O/c1-2-3-5-10-14(12-15)11-13-8-6-4-7-9-13/h4,6-9,11,15H,2-3,5,10,12H2,1H3/b14-11+

InChI key

LIPHCKNQPJXUQF-SDNWHVSQSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

α-Amylcinnamyl alcohol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under α-Amylcinnamyl alcohol


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1034512-500MG04061833839836

Questions

Reviews

No rating value

Active Filters