Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 50 mg | Check Cart for Availability | $1,480.00 |
About This Item
grade
pharmaceutical primary standard
API family
5-amino-2,4,6-triiodo-n-methylisophthalamic acid
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Ic1c(c(c(c(c1C(=O)NC)I)C(=O)O)I)N
InChI
1S/C9H7I3N2O3/c1-14-8(15)2-4(10)3(9(16)17)6(12)7(13)5(2)11/h13H2,1H3,(H,14,15)(H,16,17)
InChI key
AKXKBAWZIVNNJR-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| API family 5-amino-2,4,6-triiodo-n-methylisophthalamic acid | API family iodipamide | API family diatrizoic acid | API family iotalamic acid |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1027007-50MG | 04061833839454 |

