Skip to Content
Merck
All Photos(1)

Key Documents

Y0002263

N-Nitroso-diisopropylamine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-(1-Methylethyl)-N-nitroso-2-propanamine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

manufacturer/tradename

EDQM

concentration

in methanol

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

N(N=O)(C(C)C)C(C)C

InChI

1S/C6H14N2O/c1-5(2)8(7-9)6(3)4/h5-6H,1-4H3

InChI key

AUIKJTGFPFLMFP-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by European Pharmacopoeia. For further information and support, including product information leaflets, please go to the website of the issuing Pharmacopeia.

Application

N-Nitroso-diisopropylamine EP reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

It is also used to prepare N-Nitrosamines spiking solution to detect the concentration of N-nitrosamines in active substances according to the monograph 20542 of European Pharmacopoeia.[1]

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-NITROSAMINES IN ACTIVE SUBSTANCES
European pharmacopoeia, 20542-20542 (2022)

Articles

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

Related Content

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service