Y0002244
Aciclovir for impurity G identification
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir
About This Item
Recommended Products
API family
acyclovir
manufacturer/tradename
EDQM
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
NC1=Nc2c(ncn2COCCO)C(=O)N1
InChI
1S/C8H11N5O3/c9-8-11-6-5(7(15)12-8)10-3-13(6)4-16-2-1-14/h3,14H,1-2,4H2,(H3,9,11,12,15)
InChI key
MKUXAQIIEYXACX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service