Y0002221
Ticagrelor impurity E
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(1R,2S)-2-(3,4-Difluorophenyl)cyclopropan-1-amine (R)-mandelate
About This Item
Recommended Products
API family
ticagrelor
manufacturer/tradename
EDQM
storage temp.
2-8°C
SMILES string
Fc2c(ccc(c2)[C@H]3[C@@H](C3)[N+H3])F.[O-]C(=O)[C@H](O)c1ccccc1
InChI
1S/C9H9F2N.C8H8O3/c10-7-2-1-5(3-8(7)11)6-4-9(6)12;9-7(8(10)11)6-4-2-1-3-5-6/h1-3,6,9H,4,12H2;1-5,7,9H,(H,10,11)/t6-,9+;7-/m01/s1
InChI key
GUESUQPLVFMJIT-KLTOLQSASA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Packaging
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
425.3 °F - closed cup
Flash Point(C)
218.5 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service