Skip to Content
Merck
All Photos(1)

Key Documents

SAAR2233520

Ethyl alcohol, Pure

absolute, uniLAB

Synonym(s):

Ethanol, absolute alcohol, non-denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
MDL number:
UNSPSC Code:
12191601
Pricing and availability is not currently available.

product line

uniLAB

grade

absolute

packaging

poly bottle of 1 × 2.5 L (SAAR2233520LP)

dilution

(for laboratory use only)

storage temp.

2-30°C

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service