Skip to Content
Merck
All Photos(1)

Key Documents

PS374

Metobromuron

analytical standard

Synonym(s):

N′-(4-Bromophenyl)-N-methoxy-N-methylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BrN2O2
CAS Number:
Molecular Weight:
259.10
Beilstein:
2103964
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

manufacturer/tradename

Chem Service, Inc. PS-374

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CON(C)C(=O)Nc1ccc(Br)cc1

InChI

1S/C9H11BrN2O2/c1-12(14-2)9(13)11-8-5-3-7(10)4-6-8/h3-6H,1-2H3,(H,11,13)

InChI key

WLFDQEVORAMCIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Anić et al.
Arhiv za higijenu rada i toksikologiju, 50(2), 193-199 (1999-11-24)
An 81-year-old man was admitted in the emergency department approximately four hours after accidental, ingestion of an unknown quantity of herbicide "Galex 500 EC". This product contains 25% of metolachlor and 25% of metobromuron dissolved in xylene. In spite of
S El-Fantroussi et al.
Applied and environmental microbiology, 66(12), 5110-5115 (2000-12-01)
Soil treated with linuron for more than 10 years showed high biodegradation activity towards methoxy-methyl urea herbicides. Untreated control soil samples taken from the same location did not express any linuron degradation activity, even after 40 days of incubation. Hence
F Pank et al.
Die Pharmazie, 35(2), 115-119 (1980-02-01)
More than 40 herbicides have been tested in field trials with valerian over a period of several years. The following herbicidies have proved best suited: chloropham for application in autumn after planting, metobromurone for spraying in the early spring at
F Bonnemoy et al.
Chemosphere, 54(8), 1183-1187 (2003-12-11)
Halogenated phenylurea herbicides are not very toxic by themselves to animals, but their exposure to UV light may significantly increase the toxicity of their solutions. Absorption of light may indeed induce a phototransformation of the herbicide with a possible formation
Vincent J Smith et al.
Carbohydrate research, 344(17), 2388-2393 (2009-10-06)
We report the formation of inclusion complexes between the phenylurea herbicide metobromuron [3-(p-bromophenyl)-1-methoxy-1-methylurea] and beta- and gamma-cyclodextrin in the solid state. Formation of crystalline inclusion complexes by the kneading method was confirmed by powder X-ray diffraction and further structural characterization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service