Skip to Content
Merck
All Photos(1)

Key Documents

PS2026

Fosetyl-aluminum

technical grade

Synonym(s):

Aluminum tris(O-ethylphosphonate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HP(O)(OCH2CH3)O]3Al
CAS Number:
Molecular Weight:
354.10
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

technical grade

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-2026

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

[Al+3].CCO[PH]([O-])=O.CCO[PH]([O-])=O.CCO[PH]([O-])=O

InChI

1S/3C2H7O3P.Al/c3*1-2-5-6(3)4;/h3*6H,2H2,1H3,(H,3,4);/q;;;+3/p-3

InChI key

ZKZMJOFIHHZSRW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Fosetyl-aluminum is a fungicide, commonly used to prevent the diseases caused by Peronosporales members and foliar diseases.

Application

Fosetyl-aluminum may be used as a reference standard for the determination of fosetyl-aluminum in vegetable samples by liquid chromatography coupled with electrospray tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

No COA is supplied with Tech Grade products.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louise R Cooke et al.
Pest management science, 58(1), 17-25 (2002-02-13)
Foliar sprays of potato plants with phosphonic acid (partially neutralised with potassium hydroxide to pH 6.4) substantially reduced infection of the tubers by Phytophthora infestans, the cause of late blight, in glasshouse and field experiments over a 4-year period. Healthy
J A Quest et al.
Regulatory toxicology and pharmacology : RTP, 14(1), 3-11 (1991-08-01)
Aliette, a fungicide compound, was evaluated for carcinogenic potential by the Health Effects Division of the Office of Pesticide Programs using a consensus peer review process and EPA's guidelines for risk assessment. Aliette was categorized as a group C (possible
M López-Serrano et al.
European journal of histochemistry : EJH, 39(1), 69-74 (1995-01-01)
Grapevine (Vitis vinifera cv. Monastrell) suspension cell cultures were treated with fosetyl-A1, a widely used systemic fungicide for grapevine diseases caused by oomycetes, and examined at the electron microscope level for peroxidase cytochemistry. The results showed that treatment with fosetyl-A1
E Malusà et al.
Food additives and contaminants, 22(6), 541-548 (2005-07-16)
The levels of phosphorous acid residues in apples after foliar fertilization with P fertilizers and after treatment with a phosphonate fungicide (Fosetyl-Al) were determined and compared. Two field trials and a glasshouse experiment, using different genotypes and plants of different
Achour Amiri et al.
Phytopathology, 99(10), 1199-1208 (2009-09-11)
Isolation of Monilinia spp. from stone and pome fruit surfaces is difficult due to the presence of several fast-growing fungal species such as Rhizopus, Alternaria, and Penicillium spp. Therefore, a new selective medium (acidified potato dextrose agar [pH 3.6] amended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service