Skip to Content
Merck
All Photos(1)

Key Documents

PS1055

Aldoxycarb

analytical standard

Synonym(s):

Aldicarb-sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O4S
CAS Number:
Molecular Weight:
222.26
Beilstein:
2215242
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 10 mg

manufacturer/tradename

Chem Service, Inc. PS-1055

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(ON=CC(C)(C)S(C)(=O)=O)=O

InChI

1S/C7H14N2O4S/c1-7(2,14(4,11)12)5-9-13-6(10)8-3/h5H,1-4H3,(H,8,10)

InChI key

YRRKLBAKDXSTNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Picó et al.
Food additives and contaminants, 7 Suppl 1, S29-S34 (1990-01-01)
Aldicarb was applied to the soil of Late Valencia orange field plots. A comparative study between four analytical methods is reported, the NPD-gas chromatography method being the best choice. The work also reports the study of accumulation and persistence of
Transport of organic contaminants in groundwater: gas chromatography model to forecast the significance as applied to aldicarb sulfone residues.
J P Mestres et al.
Archives of environmental contamination and toxicology, 16(6), 649-656 (1987-11-01)
D X Lian et al.
Journal of chromatography, 542(2), 526-530 (1991-04-12)
A method is described for the determination of aldicarb and its metabolites (the sulphoxide and sulphone) in urine by gas chromatography with flame photometric detection (GC-FPD). The sample was concentrated with a column containing activated charcoal and Florisil, and then
W P Cochrane et al.
Journal - Association of Official Analytical Chemists, 64(3), 724-728 (1981-05-01)
An ion-suppression reverse phase high pressure liquid chromatographic method is described for the determination of aldicarb, aldicarb sulfoxide, and aldicarb sulfone in potatoes. Samples are extracted with methylene chloride and Na2SO4, evaporated to dryness, and cleaned up using Sep-Pak silica
Persistence and metabolism of aldicarb in fresh potatoes.
T Cairns et al.
Bulletin of environmental contamination and toxicology, 32(3), 274-281 (1984-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service