Skip to Content
Merck
All Photos(1)

Key Documents

PS102

2-Ethyl-1,3-hexanediol

analytical standard

Synonym(s):

Ethylhexylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
146.23
Beilstein:
1735324
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor density

5 (vs air)

packaging

ampule of 1000 mg

manufacturer/tradename

Chem Service, Inc. PS-102

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.451 (lit.)

bp

241-249 °C (lit.)

mp

−40 °C (lit.)

density

0.933 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CCCC(O)C(CC)CO

InChI

1S/C8H18O2/c1-3-5-8(10)7(4-2)6-9/h7-10H,3-6H2,1-2H3

InChI key

RWLALWYNXFYRGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S W Frantz et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 28(1), 1-8 (1995-11-01)
Excised skin from Fischer 344 rats, New Zealand White rabbits, and human females (obtained from mammoplasty patients) were compared for their in vitro skin penetration potential with 2-[14C]-ethyl-1,3-hexanediol (EHD). EHD was applied as both an undiluted dose and a 3%
L C Rutledge et al.
Journal of the American Mosquito Control Association, 5(3), 374-376 (1989-09-01)
The repellent ethyl hexanediol (2-ethyl-1,3-hexanediol) was tested against the mosquito Aedes dorsalis in a coastal salt marsh in California. The experimental design incorporated a linear regression model, sequential treatments and a proportional end point (95%) for protection time. The protection
S W Frantz et al.
Drug metabolism and disposition: the biological fate of chemicals, 19(5), 881-888 (1991-09-01)
A determination of the systemic pharmacokinetics of [1,3-14C]-2-ethyl-1,3-hexanediol (EHD) was conducted following i.v. dosing of male Fischer 344 rats. Pharmacokinetic analyses of the plasma data indicated that there is dose linearity in the 1.5 to 150 mg/kg range, and that
R S Slesinski et al.
Toxicology, 53(2-3), 179-198 (1988-12-30)
2-Ethyl-1,3-hexanediol (EHD) has intentional human exposure because of its application to skin as an insect repellent and its use in various skin care products. Genotoxicity studies on EHD were conducted to determine mutagenic and clastogenic potential using in vitro and
A Izri et al.
Bulletin de la Societe de pathologie exotique (1990), 94(3), 280-283 (2001-10-30)
The authors report the results of a survey on the efficacy against mosquito bites of a repellent, Mousticologne Spécial Zones Infestées (DEET 20%, EHD 15%). Two forms of the product, spray and gel, were tested in Senegal. Repellent efficacy was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service