PHR9031
2-(2,4-Difluorophenyl)-1,2,3-propanetriol
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-(2,4-Difluorophenyl)propane-1,2,3-triol
Select a Size
Select a Size
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
API family
fluconazole
product line
TraceCERT®
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
application(s)
pharmaceutical small molecule
storage temp.
2-8°C
SMILES string
OCC(C1=CC=C(F)C=C1F)(O)CO
InChI
1S/C9H10F2O3/c10-6-1-2-7(8(11)3-6)9(14,4-12)5-13/h1-3,12-14H,4-5H2
InChI key
ZHVKNFSTIAMHSM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com
Application
Other Notes
Recommended products
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service