Skip to Content
Merck
All Photos(1)

Key Documents

PHL89593

Camptothecin

phyproof® Reference Substance

Synonym(s):

(S)-(+)-Camptothecin

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$415.00

$415.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$415.00

About This Item

Empirical Formula (Hill Notation):
C20H16N2O4
CAS Number:
Molecular Weight:
348.35
Beilstein:
631069
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$415.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

mp

260 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5ccccc5nc34)C2=O

InChI

1S/C20H16N2O4/c1-2-20(25)14-8-16-17-12(7-11-5-3-4-6-15(11)21-17)9-22(16)18(23)13(14)10-26-19(20)24/h3-8,25H,2,9-10H2,1H3/t20-/m0/s1

InChI key

VSJKWCGYPAHWDS-FQEVSTJZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application




  • Co-delivery system for camptothecin and doxorubicin: Research on dendritic polymer prodrug-based unimolecular micelles demonstrated a pH-responsive co-delivery mechanism for camptothecin and doxorubicin, offering a synergistic effect in controlled drug release (Chen and Liu, 2024).


  • Investigation into camptothecin′s role in chronic myeloid leukemia: The study explored the therapeutic potential of FL118, a camptothecin derivative, against chronic myeloid leukemia resistant to BCR-ABL inhibitors, targeting RNA helicase DDX5 (Takeda et al., 2024).


Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raju Suresh Kumar et al.
Saudi journal of biological sciences, 27(12), 3290-3300 (2020-12-12)
A small library of cage-like heterocyclic hybrids encompassing pyrroloisoquinolines, pyridinone and acenaphthene structural moieties have been synthesized and tested for their potential as anticancer agents against HCT116 and JURKAT cell lines. The results revealed that these cell lines are more
Katerina D Fagan-Solis et al.
Cell reports, 30(5), 1385-1399 (2020-02-06)
The Mre11-Rad50-Nbs1 complex is a DNA double-strand break sensor that mediates a tumor-suppressive DNA damage response (DDR) in cells undergoing oncogenic stress, yet the mechanisms underlying this effect are poorly understood. Using a genetically inducible primary mammary epithelial cell model
Jun Ho Ko et al.
Cell reports, 33(12), 108543-108543 (2020-12-29)
DNA damage tolerance (DDT) and homologous recombination (HR) stabilize replication forks (RFs). RAD18/UBC13/three prime repair exonuclease 2 (TREX2)-mediated proliferating cell nuclear antigen (PCNA) ubiquitination is central to DDT, an error-prone lesion bypass pathway. RAD51 is the recombinase for HR. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service