PHL89217
Glycyrrhizin
phyproof® Reference Substance
Synonym(s):
Glycyrrhizic acid, 3-O-(2-O-β-D-Glucopyranuronosyl-α-D-glucopyranuronosyl)-18β-glycyrrhetinic acid, Glycyrrhetinic acid glycoside, Glycyrrhizinic acid
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
storage temp.
2-8°C
SMILES string
C[C@]1(C(O)=O)CC[C@@](CC[C@]([C@@]2(C)[C@@]3([H])[C@]4(C)[C@@](C(C)(C)[C@@H](O[C@@]5([H])O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]5O[C@@]6([H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O6)CC4)([H])CC2)(C)C7=CC3=O)(C)[C@@]7([H])C1
InChI
1S/C42H62O16/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54)/t19-,21-,22-,23-,24-,25-,26-,27+,28-,29-,30+,31+,34-,35-,38+,39-,40-,41+,42+/m0/s1
InChI key
LPLVUJXQOOQHMX-QWBHMCJMSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service