Skip to Content
Merck
All Photos(1)

Key Documents

PHL85752

Isoscopoletin

phyproof® Reference Substance

Synonym(s):

6-Hydroxy-7-methoxycoumarin, 7-Methoxyesculetin, Esculetin 7-methyl ether, Isoscopoletin

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$427.00

$427.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$427.00

About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
Beilstein:
157280
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$427.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

SMILES string

COc1cc2OC(=O)C=Cc2cc1O

InChI

1S/C10H8O4/c1-13-9-5-8-6(4-7(9)11)2-3-10(12)14-8/h2-5,11H,1H3

InChI key

SYTYLPHCLSSCOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Isoscopoletin is a coumarin that is produced in many plant species by the demethylation of scoparone. It inhibits proliferation of both control and multidrug resistant leukemia cell lines. It also inhibits proliferation of hepatitis B virus in cellular systems.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service