Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Check Cart for Availability | $298.00 |
About This Item
Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
127683
MDL number:
grade
primary reference standard
product line
phyproof® Reference Substance
assay
≥95.0% (HPLC)
form
crystals
manufacturer/tradename
PhytoLab
mp
230 °C (dec.) (lit.)
SMILES string
Oc1ccc2C=CC(=O)Oc2c1
InChI
1S/C9H6O3/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5,10H
InChI key
ORHBXUUXSCNDEV-UHFFFAOYSA-N
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Other Notes
This compound is commonly found in plants of the genus: angelica glycyrrhiza pimpinella
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
1 of 1
This Item | |||
|---|---|---|---|
| grade primary reference standard | grade analytical standard | grade - | grade - |
| mp 230 °C (dec.) (lit.) | mp 230 °C (dec.) (lit.) | mp 230 °C (dec.) (lit.), 230-234 °C | mp 230 °C (dec.) (lit.) |
| assay ≥95.0% (HPLC) | assay ≥98.0% (HPLC) | assay ≥98.0% (HPLC) | assay 99% |
| form crystals | form - | form crystals | form - |
| manufacturer/tradename PhytoLab | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename - |
| product line phyproof® Reference Substance | product line - | product line - | product line - |
Still not finding the right product?
Explore all of our products under Umbelliferone
Storage Class
11 - Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
