Skip to Content
Merck
All Photos(1)

Key Documents

N11793

Diphacinone

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-288

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

InChI

1S/C23H16O3.Na/c24-21-17-13-7-8-14-18(17)22(25)20(21)23(26)19(15-9-3-1-4-10-15)16-11-5-2-6-12-16;/h1-14,19-20H;/q;+1

InChI key

YCKTVRYJMHFIRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 4 Inhalation - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barnett A Rattner et al.
Ecotoxicology and environmental safety, 73(6), 1159-1164 (2010-06-29)
The anticoagulant rodenticide diphacinone was slightly toxic (acute oral LD50 2014 mg/kg) to Northern bobwhite (Colinus virginianus) in a 14-day acute toxicity trial. Precise and sensitive assays of blood clotting (prothrombin time, Russell's Viper venom time, and thrombin clotting time)
M E Mount et al.
American journal of veterinary research, 44(11), 2009-2017 (1983-11-01)
Vitamin K1 (5 mg/kg of body weight/day divided for several 5-day regimens) was effective in preventing bleeding diathesis in diphacinone-poisoned dogs. Diphacinone, a vitamin K-inhibiting rodenticide, was given 2.5 mg of diphacinone/kg of body weight orally in divided doses 2
Thomas Primus et al.
Journal of chromatographic science, 44(1), 1-5 (2006-04-08)
A reversed-phase ion-pair liquid chromatographic analysis combined with a solid-phase extraction clean-up method is used to assess the quantity of diphacinone residue found in invertebrates. Three invertebrate species are exposed to commercially available diphacinone-fortified bait used for rat control. The
Diphacinone toxicity, von Willebrand's Disease, and Ehrlichia canis in a dog.
G C Troy
The Veterinary clinics of North America. Small animal practice, 18(1), 255-257 (1988-01-01)
Henry A Spiller et al.
Veterinary and human toxicology, 45(6), 313-314 (2003-12-03)
Rare cases of coagulaopathies from dermal absorption of hydroxycoumarin derivatives have been reported. We report the first case of dermal absorption of an indandione derivative rodenticide causing severe coagulopathy. An 18-y-o male worker at a pest exterminating company spilled a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service