Skip to Content
Merck
All Photos(1)

Key Documents

N11454

Chlorothalonil

analytical standard

Synonym(s):

Tetrachloroisophthalodinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8Cl4N2
CAS Number:
Molecular Weight:
265.91
Beilstein:
1978326
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-1020

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Clc1c(Cl)c(C#N)c(Cl)c(C#N)c1Cl

InChI

1S/C8Cl4N2/c9-5-3(1-13)6(10)8(12)7(11)4(5)2-14

InChI key

CRQQGFGUEAVUIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H S Rosenkranz
Mutation research, 423(1-2), 183-186 (1999-02-25)
Based upon analyses using a number of validated structure-activity relationship models, it is concluded that the carcinogenicity in rodents of chlorothalonil is not due to a genotoxic mechanism.
Shuangying Yu et al.
Environmental pollution (Barking, Essex : 1987), 181, 329-334 (2013-07-23)
Chlorothalonil is a broad spectrum fungicide widely used in agricultural and urban environments, yet little is known regarding its effects on amphibians. We examined effects of chlorothalonil on growth, malformations, and mortality in embryos and larvae of Xenopus laevis and
Taegan A McMahon et al.
Ecology letters, 15(7), 714-722 (2012-05-17)
Although studies on biodiversity and ecosystem function are often framed within the context of anthropogenic change, a central question that remains is how important are direct vs. indirect (via changes in biodiversity) effects of anthropogenic stressors on ecosystem functions in
Marie E DeLorenzo et al.
Marine pollution bulletin, 64(7), 1291-1299 (2012-06-12)
The precise application of risk assessment can lead to different conclusions about risk depending on how species are grouped in the assessment. We compared the use of different risk assessment methods for three different classes of pesticide, the herbicide diuron
Timothy J Reilly et al.
Chemosphere, 89(3), 228-234 (2012-05-09)
To provide an assessment of the occurrence of fungicides in water resources, the US Geological Survey used a newly developed analytical method to measure 33 fungicides and an additional 57 current-use pesticides in water samples from streams, ponds, and shallow

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service