MSCAL5-1EA
ProteoMass™ LTQ/FT-Hybrid ESI Pos. Mode Cal Mix
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
12142208
Recommended Products
product line
ProteoMass™
packaging
pkg of 1 × 10 mL
concentration
(varied conc.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
The ProteoMass ESI Calibration Kit for LTQ based Hybrids, positive ion mode calibration solution, enables calibration of Thermo Scientific hybrid instruments equipped with ESI source in positive ion mode and covers a mass range from m/z 138 to m/z 1822 and is therefore useable for calibrations between m/z 50 and 2000.
Features and Benefits
Discover the Advantages for Yourself!
- Ready-to-use
- Developed and qualified specifically for use with the LTQ based Thermo Scientific hybrid instruments (LTQ FT, LTQ FT Ultra, and LTQ Orbitrap series) with ESI source and Thermo Scientific Exactive
- Conveniently packaged, freeing you from time consuming mixing and dilution steps, allowing you to focus on acquisition of quality data
Legal Information
ProteoMass is a trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
ProteoMass is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Fang Chen et al.
Proteome science, 14, 17-17 (2016-11-30)
Human follicular fluid (FF) is a unique biological fluid in which the oocyte develops in vivo, and presents an optimal source for non-invasive biochemical predictors. Oocyte quality directly influences the embryo development and hence, may be used as a predictor
Christoph-Martin Geilfus et al.
Journal of proteomics, 169, 33-40 (2017-06-20)
Phosphorus is often the least available macronutrient in soil. Lack in phosphorus has detrimental effect on growth and biomass production of European Fagus sylvatica L., a major trees species in temperate forests. In contrast to leaf tissues, few studies have
Marcus Dittrich et al.
Nature plants, 5(9), 1002-1011 (2019-08-28)
Stomata are microscopic pores found on the surfaces of leaves that act to control CO2 uptake and water loss. By integrating information derived from endogenous signals with cues from the surrounding environment, the guard cells, which surround the pore, 'set'
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service