K5112-U
Omnifit® EZ Benchmark Columns with 1 Adjustable and 1 fixed Endpiece
I.D. × L 15 mm × 100 mm
Synonym(s):
Omnifit column, 10 mm x 150 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Omnifit 006EZ-15-10-AF
Quality Level
technique(s)
LPLC: suitable
I.D. × L
15 mm × 100 mm
Looking for similar products? Visit Product Comparison Guide
General description
Omnifit® Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system. Two configurations are available:
- Non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.
- One fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80mm of height adjustment.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service