Skip to Content
Merck
All Photos(1)

Key Documents

ERMCZ110

Fine Dust (PM2,5 like) (extractable ions)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$376.00

$376.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$376.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$376.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

Agency

ERM®

manufacturer/tradename

JRC

application(s)

environmental

storage temp.

2-8°C

Analysis Note

For more information please click on the below product number:
ERMCZ110

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service