Skip to Content
Merck
All Photos(1)

Documents

ERMCE102

Fish Tissue (PBDEs in fish tissue)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

ERM®

manufacturer/tradename

JRC

application(s)

food and beverages

storage temp.

2-8°C

Other Notes

Certified for the analytes listed below. See certificate for values and more details

Organic Pollutants:
BDE-47 (2,2′,4,4′-tetrabromodiphenyl ether)
BDE-49 (2,2′,4,5′-tetrabromodiphenyl ether)
BDE-99 (2,2′,4,4′,5-pentabromodiphenyl ether)
BDE-100 (2,2′,4,4′,6-pentabromodiphenyl ether)
BDE-153 (2,2′,4,4′,5,5′-hexabromodiphenyl ether)
BDE-154 (2,2′,4,4′,5,6′-hexabromodiphenyl ether)

Indicative Values:
BDE-28 (2,4,4′-tribromodiphenyl ether)
BDE-183 (2,2′,3,4,4′,5′,6-heptabromodiphenyl ether)

Matrix Group: Fish and Seafood

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service