Skip to Content
Merck
All Photos(1)

Key Documents

CRM80876

Supelco

Pinoxaden Metabolite SYN502836

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,5-Diethyl-4-(hexahydro-7,9-dioxo-7H-pyrazolo[1,2-d][1,4,5]oxadiazepin-8-yl)-benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$189.00

$189.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$189.00

About This Item

Empirical Formula (Hill Notation):
C18H22N2O5
CAS Number:
Molecular Weight:
346.38
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$189.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

N21N(CCOCC2)C(=O)C(C1=O)c3c(cc(cc3CC)C(=O)O)CC

InChI

1S/C18H22N2O5/c1-3-11-9-13(18(23)24)10-12(4-2)14(11)15-16(21)19-5-7-25-8-6-20(19)17(15)22/h9-10,15H,3-8H2,1-2H3,(H,23,24)

InChI key

PZURTZWDEUDQOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Pinoxaden Metabolite SYN502836 is formed as a metabolite of the new class of phenylpyrazoline herbicides, pinoxaden in plants. It impedes the activity of acetyl CoA carboxylase (ACCase) and prevents fatty acid biosynthesis.
As per Regulation (EC) No. 1107/2009, repealing directive 91/414/EEC, pinoxaden is approved for use in the European Union (EU) as a plant protection agent.
The EU has proposed to change the residue definition of pinoxaden in the draft regulation SANTE/10776/2021. Based on the results of a metabolism study conducted in primary and rotational crops, the residue definition of pinoxaden for risk assessment is now proposed to be measured as a sum of pinoxaden metabolites ― M4 (SYN 505164) and M6 (SYN 502836), in both free and conjugated forms.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Resp. Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service