CRM69184
4-Phenylphenol
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
4-Hydroxybiphenyl
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
bp
321 °C (lit.)
mp
164-166 °C (lit.)
format
neat
SMILES string
Oc1ccc(cc1)-c2ccccc2
InChI
1S/C12H10O/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9,13H
InChI key
YXVFYQXJAXKLAK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
4-Phenylphenol CRM can also be used as follows:
- Multi-residue analysis of 24 endocrine-disrupting chemicals (EDCs) from six different chemical families in 10 fish and seafood samples by combining ultrasound-assisted extraction with solid-phase extraction (SPE) followed by gas chromatographic-mass spectrometric (GC-MS) determination
- Simultaneous determination of six varieties of disrupting chemicals (EDCs) in 13 cereal-based food products by ultrasound-assisted extraction-solid phase extraction-gas chromatography-mass spectrometry (UAE-SPE-GC-MS)
- Evaluation of endocrine-disrupting chemicals in 11 fresh vegetable samples and 7 fruit samples by ultrasound-assisted extraction (UAE) followed by solid-phase extraction (SPE) and gas chromatography-mass spectrometry (GC-MS) detection
- Development and validation of a gas chromatography-mass spectrometry (GC-MS) based method following microwave-assisted derivatization for quantifying 23 EDCs in 33 samples of dairy products
- Multi-residue determination of 18 preservatives in different vegetable samples by combining dispersive-solid phase extraction (d-SPE) with ultra-high performance liquid chromatography-triple quadrupole/linear ion trap mass spectrometry (UHPLC-QTRAP)
Recommended products
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1B
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
320.0 °F - closed cup
Flash Point(C)
160 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service