Skip to Content
Merck
All Photos(1)

Key Documents

CRM69056

Supelco

Disulfoton-sulfone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$200.00

$200.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$200.00

About This Item

Empirical Formula (Hill Notation):
C8H19O4PS3
CAS Number:
Molecular Weight:
306.40
Beilstein:
1713538
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$200.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SCCS(=O)(=O)CC

InChI

1S/C8H19O4PS3/c1-4-11-13(14,12-5-2)15-7-8-16(9,10)6-3/h4-8H2,1-3H3

InChI key

BKVJOVPVLOJPKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Disulfoton-sulfone is an oxidative metabolite of the organophosphorus insecticide disulfoton.
The European Union regulations define a maximum residue limit (MRL) of 0.01 mg/kg for disulfoton in foods of plant origin which is taken as a sum of disulfoton, disulfoton sulfoxide, and disulfoton sulfone. Currently, disulfoton is not allowed for its use in the European Union and also has restrictions over its use in the USA by American Environmental Protection Agency (EPA).

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The disulfoton-sulfone CRM may also find given below uses:
  • Determination of disulfoton and its five metabolites in human blood and urine samples by QuEChERS extraction followed by liquid chromatography-tandem mass spectrometric analysis
  • Method development for the analysis of 15 organophosphorus pesticides and their metabolites in water samples using multiwalled carbon nanotubes based dispersive solid phase extraction and gas chromatography-nitrogen phosphorus detector (GC-NPD)
  • Simultaneous determination of 15 organophosphorus pesticides and their metabolites in cereal-based baby food samples using multi-walled carbon nanotubes based dispersive solid phase extraction and gas chromatography-nitrogen phosphorus detector (GC-NPD)
  • Analysis of 11 pesticides and 4 of their metabolites in chickpeas and barley flours using a modified QuEChERS extraction and GC-NPD determination
  • Multi-residue determination of 60 pesticides in hen egg samples by liquid-chromatography and gas chromatography with tandem mass spectrometry, in combination with QuEChERS based sample pre-treatment
  • Method development to quantify 49 organophosphorus pesticide residues and 10 of their metabolites in fish, egg, and milk samples by means of dual gas chromatography-dual pulse flame photometric detection

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Ángel González-Curbelo et al.
Journal of separation science, 36(3), 556-563 (2013-01-11)
In this manuscript, a dispersive SPE method based on the use of multiwalled carbon nanotubes has been developed for the determination of 15 organophosphorus pesticides residues including some of their metabolites (disulfoton sulfoxide, ethoprophos, cadusafos, dimethoate, terbufos, disulfoton, chlorpyrifos-methyl, malaoxon
Lixin Yang et al.
Journal of agricultural and food chemistry, 60(8), 1906-1913 (2012-02-04)
A new method for the quantitative determination of 49 kinds of organophosphorus pesticide residues and their metabolites in fish, egg, and milk by dual gas chromatography-dual pulse flame photometric detection was developed. Homogenized samples were extracted with acetone and methylene
Miguel Á González-Curbelo et al.
Journal of separation science, 35(2), 299-307 (2012-01-01)
Analytical potentiality of a modified version of the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) method has been studied and validated for the extraction of a group of 11 pesticides (ethoprophos, cadusafos, dimethoate, terbufos, disulfoton, chlorpyrifos-methyl, fenitrothion, pirimiphos-methyl, malathion
Kiyotaka Usui et al.
Legal medicine (Tokyo, Japan), 14(6), 309-316 (2012-07-24)
A liquid chromatography-tandem mass spectrometry method was developed and validated for simultaneous determination of disulfoton and five of its oxidative metabolites (disulfoton-sulfoxide, disulfoton-sulfone, demeton-S, demeton-S-sulfoxide and demeton-S-sulfone) in human whole blood and urine. Extraction was undertaken using a QuEChERS method
Miguel Angel González-Curbelo et al.
Analytical and bioanalytical chemistry, 404(1), 183-196 (2012-05-25)
In the present study, a new analytical method has been developed for the simultaneous quantification of 15 organophosphorus pesticides, including some of their metabolites, (disulfoton-sulfoxide, ethoprophos, cadusafos, dimethoate, terbufos, disulfoton, chlorpyrifos-methyl, malaoxon, fenitrothion, pirimiphos-methyl, malathion, chlorpyrifos, terbufos-sulfone, disulfoton-sulfone and fensulfothion)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service