Skip to Content
Merck
All Photos(1)

Key Documents

CRM48905

Supelco

Polynuclear Aromatic Hydrocarbons Mix

certified reference material, TraceCERT®, 2000 μg/mL each component in methylene chloride: benzene (1:1)

Synonym(s):

PNAs

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$185.00

$185.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$185.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$185.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methylene chloride: benzene (1:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Anthracene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Chrysene
    Dibenz[a,h]anthracene
    Fluoranthene
    Fluorene
    Indeno[1,2,3-cd]pyrene
    Naphthalene
    Phenanthrene
    Pyrene
    See All (16)

Signal Word

Danger

Flash Point(F)

12.0 °F

Flash Point(C)

-11.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abhrajyoti Tarafdar et al.
Environmental geochemistry and health, 42(6), 1691-1704 (2019-05-28)
Children can get affected by polycyclic aromatic hydrocarbons (PAHs) while they interact with play area soil/rubber surfacing and exposed to PAHs by dermal contact, inhalation and hand-to-mouth activity. A comparative study has been conducted on PAHs profiling and probable cancer
José Beirão et al.
Environmental pollution (Barking, Essex : 1987), 241, 521-528 (2018-06-09)
The effects of petroleum aromatic hydrocarbons (PAHs) on the embryonic and larval life stages of teleosts have been extensively examined. However, very little work has been conducted on how spilled oil affects fish sperm and there is no related knowledge
St Naydenova et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 55(4), 498-509 (2019-12-19)
A GC-MS method in SIM mode is proposed for routine analysis of nineteen polycyclic aromatic hydrocarbons (PAHs) in particulate matter with an aerodynamic diameter of 10 µm. The latter is collected on Whatman® QM-A quartz filters via OPSIS SM200 sampler during

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service