Skip to Content
Merck
All Photos(1)

Key Documents

CRM18501

Supelco

Ametoctradin hydrochloride Metabolite M650F06

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

6-(7-Amino-5-ethyl[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)hexanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$121.00

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$121.00

About This Item

Empirical Formula (Hill Notation):
C13H19N5O2· HCl
CAS Number:
Molecular Weight:
313.78
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

Cl.[n]21ncnc2NC(=CC)C(=C1N)CCCCCC(=O)O

InChI key

XJCSIYKKYXQBGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

M650F06 is a metabolite of ametoctradin.
Ametoctradin is a contact fungicide with protective action and is used for controlling oomycetes such as Plasmopara viticola in grapes, Phytophthora infestans in potatoes and tomatoes, Phytophthora, Peronospora, and Pseudoperonospora spp. in vegetables. EFSA established a provisional residue definition for enforcement and risk assessment in animal matrices as “ametoctradin, M650F01, and M650F06, expressed as ametoctradin”.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service