Skip to Content
Merck
All Photos(1)

Documents

CRM04291

Supelco

Diclofop-methyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14Cl2O4
CAS Number:
Molecular Weight:
341.19
Beilstein:
2224754
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

SMILES string

COC(=O)C(C)Oc1ccc(Oc2ccc(Cl)cc2Cl)cc1

InChI

1S/C16H14Cl2O4/c1-10(16(19)20-2)21-12-4-6-13(7-5-12)22-15-8-3-11(17)9-14(15)18/h3-10H,1-2H3

InChI key

BACHBFVBHLGWSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Diclofop-methyl is a post-emergence chiral herbicide that belongs to the class of aryloxyphenoxy propanoate (AOPP) compounds. It prevents the synthesis of fatty acids by inhibiting the activity of acetyl CoA carboxylase (ACCase). Diclofop-methyl is a selective and systemic herbicide used against wild oats, wild millets, and other annual grass weeds in broad-leaf crops, wheat, barley, maize, sorghum, oats, sugar cane, rice, dicotyledonous vegetables, and cotton. It causes an immediate inhibitory effect on the growth of the shoot, intercalary, and root meristems of the plant. It quickly hydrolyzes to give diclofop in plants, water, and soil.
Diclofop-methyl was included on 1st June 2011 in Annex I of Directive 91/414/EEC by the European Commission Directive 2011/45/EU. Diclofop-methyl is approved for use in the European Union under EC Regulation No 1107/2009, as per the Commission Implementing Regulation (EU) No 540/2011, however it is a candidate for substitution.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The diclofop-methyl CRM may find its use as described below:

  • To study the enantioselective degradation of diclofop-methyl and diclofop in two soil samples under aerobic and anaerobic conditions using high-performance liquid chromatography (HPLC)
  • Simultaneous extraction and determination of enantiomers of diclofop-methyl and diclofop in cole using high-performance liquid chromatography-chiral stationary phase (HPLC-CS)
  • Development of a multi-residue method for simultaneous estimation of 16 pesticides in water samples using direct immersion SPME followed by GC-MS analysis
  • Simultaneous extraction and determination of aryloxyphenoxy-propionate herbicides from water samples by dispersive magnetic solid-phase extraction (d-MSPE) with high-performance liquid chromatography-diode array detector (HPLC-DAD) and ultra-high pressure liquid chromatography -triple quadrupole mass spectrometer (UHPLC-MS/MS)
  • Evaluating the capacity of silica-bonded deoxycholic acid as a stationary phase having a 3,5-dinitrophenylcarbamoyl and a calix[4]arene substituent for enantiomeric separations

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective degradation of diclofop-methyl in cole (Brassica chinensis L.)
Gu X, et al.
Food Chemistry, 121(1), 264-267 (2010)
Maciej Tankiewicz et al.
Talanta, 107, 1-10 (2013-04-20)
A simple and solvent-free multi-residue method has been optimized to determine 16 currently used pesticides from different chemical groups in aqueous samples. The extraction of analytes was carried out with direct immersion solid-phase microextraction (DI-SPME) and for the identification and
Sadegh Yaghoubnejad et al.
Journal of separation science, 41(9), 1903-1912 (2018-01-16)
We report the synthesis and enantioseparation characteristics of two novel covalently immobilized deoxycholic acid derivatives as chiral stationary phases for high-performance liquid chromatography. In the structure of the first stationary phase, the 3-position of deoxycholic acid is substituted with a
Jinling Diao et al.
Environmental science & technology, 44(6), 2042-2047 (2010-02-17)
In this study, the degradation of diclofop-methyl (DM) and its main metabolite, diclofop (DC), in two soils under aerobic and anaerobic conditions were investigated using enantioselective HPLC. Under aerobic or anaerobic conditions, rapid hydrolysis to the corresponding acid diclofop (DC)was
Review of the existing maximum residue levels for diclofop (considered variant diclofop-methyl) according to Article 12 of Regulation (EC) No 396/2005
European Food Safety Authority (EFSA), et al.
EFSA Journal, 18(1), e05981-e05981 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service