Skip to Content
Merck
All Photos(1)

Key Documents

CRM01399

Supelco

Teflubenzuron

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Clofentezine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$154.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$154.00

About This Item

Empirical Formula (Hill Notation):
C14H6Cl2F4N2O2
CAS Number:
Molecular Weight:
381.11
Beilstein:
8229925
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$154.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

Fc1cccc(F)c1C(=O)NC(=O)Nc2cc(Cl)c(F)c(Cl)c2F

InChI

1S/C14H6Cl2F4N2O2/c15-5-4-8(12(20)10(16)11(5)19)21-14(24)22-13(23)9-6(17)2-1-3-7(9)18/h1-4H,(H2,21,22,23,24)

InChI key

CJDWRQLODFKPEL-UHFFFAOYSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Teflubenzuron is a type of benzoylurea insecticide that disrupts the chitin deposition in the insect cuticle by inhibiting chitin synthesis and molting. It is a systemic insect growth regulator used for the control of lepidoptera, coleoptera, diptera, aleyrodidae, hymenoptera, psyllidae, and hemiptera larvae in vines, pome fruit, stone fruit, citrus, cabbages, potatoes, vegetables, soybeans, trees, sorghum, tobacco, and cotton. It is also used to control fly and mosquito larvae and immature stages of locusts.
Teflubenzuron was included to the Annex I of Directive 91/414/EEC on 01 December 2009. Teflubenzuron is not approved for use in the European Union (EU). But the Annexe IIIA of Regulation (EC) No 396/2005 establishes the maximum residue levels (MRLs) of teflubenzuron in the range of 0.01–5 mg/kg in fruits, vegetables, crops, and livestock. Teflubenzuron is monitored in the Multiannual Control Programmes for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques.
The teflubenzuron certified reference material (CRM) may also find use as given below:
  • Determination of 9 benzoylurea insecticides in fruit, vegetable, cereal, and animal product samples by pressurized liquid extraction and LC-MS/MS
  • Simultaneous analysis of lufenuron, bifenthrin, and teflubenzuron residues in coconut water by matrix solid-phase dispersion extraction combined with reversed-phase high-performance liquid chromatography
  • Development of a high performance liquid chromatography–triple quadrupole mass spectrometry (HPLC-MS/MS) based method for the simultaneous estimation of 7 benzoylurea insecticides in infused oolong tea
  • Multi-residue analysis of pesticides in Açai fruit by an extraction based on matrix solid phase dispersion and determination using liquid chromatography
  • Comparative study for the extraction of 9 benzoylurea insecticides using two dispersive liquid-liquid microextraction (DLLME) approaches from wastewater samples followed by their determination using liquid chromatography-quadrupole-linear ion trap-mass spectrometry

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Chen et al.
Food chemistry, 143, 405-410 (2013-09-24)
A method for the simultaneous determination of 7 benzoylurea pesticides (chlorfluazuron, diflubenzuron, fluazuron, flufenoxuron, hexaflumuron, teflubenzuron and triflumuron) in the manufactured Oolong tea leaves and its infusion was described. The method has a LOD of 0.03-1.00ng/mL, a recovery of 90.4-103%
Multi-residue determination of pesticides in acai tropical fruit (Euterpe oleracea) by matrix solid-phase dispersion combined with liquid chromatography
Fr?es, M B R, et al.
Food Analytical Methods, 328-333 null
Monia Brutti et al.
Journal of separation science, 33(1), 1-10 (2010-01-22)
A method based on pressurized liquid extraction and LC-MS/MS has been developed for determining nine benzoylureas (BUs) in fruit, vegetable, cereals, and animal products. Samples (5 g) were homogenized with diatomaceous earth and extracted in a 22 mL cell with
Reasoned opinion on the review of the existing maximum residue levels (MRLs) for teflubenzuron according to Article 12 of Regulation (EC) No 396/2005
European Food Safety Authority
EFSA Journal, 3664 null
A reversed-phase high-performance liquid chromatography method combined with matrix solid-phase dispersion extraction for the determination of teflubenzuron, lufenuron and bifenthrin residues in lyophilized coconut water
Santos L F S, et al.
Journal of food composition and analysis, 183-188 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service