Skip to Content
Merck
All Photos(1)

Key Documents

93032

Azadirachtin

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

93032-50MG
$1,290.00

$1,290.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
93032-50MG
$1,290.00

About This Item

Empirical Formula (Hill Notation):
C35H44O16
CAS Number:
Molecular Weight:
720.71
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,290.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

[H]C1([H])C2OC3([H])OC=CC3(O)C1C4(C)OC24[C@@]5(C)[C@@H](O)[C@@H]6OC[C@]7([C@H](C[C@H](OC(=O)C(C)=CC)[C@]8(COC(O)([C@H]58)C(=O)OC)[C@@]67[H])OC(C)=O)C(=O)OC

InChI

1S/C35H44O16/c1-8-15(2)24(38)49-18-12-19(48-16(3)36)32(26(39)43-6)13-46-21-22(32)31(18)14-47-34(42,27(40)44-7)25(31)29(4,23(21)37)35-20-11-17(30(35,5)51-35)33(41)9-10-45-28(33)50-20/h8-10,17-23,25,28,37,41-42H,11-14H2,1-7H3/t17?,18-,19?,20?,21+,22+,23?,25?,28?,29?,30?,31?,32-,33?,34-,35?/m0/s1

InChI key

FTNJWQUOZFUQQJ-CFYAVYSVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Biochem/physiol Actions

Triterpenoid found in need tree seeds, azadiractin suppresses feeding by many insect species and disrupts growth of most insect and other arthropod species, while having very low mammalian toxicity. Promising as a natural pesticide.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service