Skip to Content
Merck
All Photos(1)

Key Documents

89453

Supelco

Isoflucypram

PESTANAL®, analytical standard

Synonym(s):

N-[[5-Chloro-2-(1-methylethyl)phenyl]methyl]-N-cyclopropyl-3-(difluoromethyl)-5-fluoro-1-methyl-1H-pyrazole-4-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$102.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$102.00

About This Item

Empirical Formula (Hill Notation):
C19H21ClF3N3O
CAS Number:
Molecular Weight:
399.84
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water (calc. from elemental analysis)

mp

106-111 °C

format

neat

SMILES string

CC(C1=C(CN(C(C2=C(N(N=C2C(F)F)C)F)=O)C3CC3)C=C(Cl)C=C1)C

InChI

1S/C19H21ClF3N3O/c1-10(2)14-7-4-12(20)8-11(14)9-26(13-5-6-13)19(27)15-16(17(21)22)24-25(3)18(15)23/h4,7-8,10,13,17H,5-6,9H2,1-3H3

InChI key

JEFUQUGZXLEHLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isoflucypram, termed as a new succinate dehydrogenase inhibitor (SDHI) fungicide, is a tertiary amine, containing a cyclopentyl group at the N-atom in its structural orientation. It is found to be effective in controlling rusts, leaf spot, powdery mildew, sclerotinia, septoria tritici and botrytis cereal diseases.[1]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frontiers Of Organofluorine Chemistry (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service