Skip to Content
Merck
All Photos(1)

Key Documents

845106

TOYOPEARL GigaCap® DEAE-650 MiniChrom Column

phase DEAE (diethylaminoethyl), 75 μm particle size, pore size 1000 Å, L × I.D. 10 cm × 8 mm, MiniChrom Column

Synonym(s):

MiniChrom

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$347.00

$347.00


Estimated to ship onMay 15, 2025



Select a Size

Change View
1 EA
$347.00

About This Item

UNSPSC Code:
41106500

$347.00


Estimated to ship onMay 15, 2025


material

PEEK

product line

TOYOPEARL®

feature

counter ion Cl-

analyte chemical class(es)

proteins

manufacturer/tradename

Tosoh 45106

parameter

3 bar max. pressure (resin)
30 bar max. pressure (column)

technique(s)

HPLC: suitable
LPLC: suitable

L × I.D.

10 cm × 8 mm

bed volume

5 mL

matrix

hydroxylated methacrylic polymer

matrix active group

DEAE (diethylaminoethyl) phase

particle size

75 μm

pore size

1000 Å

operating pH

2-13

capacity

>156 g/L adsorption capacity (BSA)

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

MiniChrom columns for TOYOPEARL® and TSKgel® are pre-packed with the most popular ion exchange, mixed-mode, hydrophobic interaction or affinity media. These columns are available in 5 mL volume, 8 mm ID x 10 cm L, and can be operated with standard laboratory LC systems. MiniChrom columns for TOYOPEARL and TSKgel provide:
  • Direct connection to common laboratory LC Systems
  • Easy parameter screening and method optimization
  • Fast small scale purification
  • Reliable and reproducible results

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol, 0.15 M NaCl.

Other Notes

Columns are tolerant to aqueous buffers and salt solutions, 1 mol/L alkaline solutions, 1 mol/L non-oxidizing mineral acids, 8 mol/L urea, 6 mol/L guanidine hydrochloride, non-halogenated organic solvents and detergents. They are not compatible with strong oxidants and halogenated solvents.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation
Toyopearl is a registered trademark of Tosoh Corporation
Toyopearl GigaCap is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service