Skip to Content
Merck
All Photos(1)

Key Documents

822806

TOYOPEARL® AF-rProtein A-650F Bulk Media

phase rProtein A, bottle of 1 L, 45 μm particle size

Synonym(s):

TOYOPEARL® AF-rProtein A-650F Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
Pricing and availability is not currently available.

description

Adsorption capacity (> 45 g/L IgG (static); >30 g/L IgG (dynamic at 2 min. residence time))
Clean in Place (CIP) (0.1 M NaOH)
Phase filter (Protein A)
Shipping solvent (20% (v/v) ethanol)

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 22806

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

rProtein A phase

particle size

45 μm

pore size

1000 Å

pH

2-13

capacity

>30 g/L adsorption capacity (IgG)(dynamic at 2 min. residence time)
>45 g/L adsorption capacity (IgG)(static)

separation technique

affinity

Looking for similar products? Visit Product Comparison Guide

General description

Protein A chromatography, the most widely used type of affinity chromatography, relies on the specific and reversible binding of antibodies to an immobilized ligand; in this case protein A. Protein A is a 56 kDa surface protein native to the cell wall of the bacterium Staphylococcus aureus. It is composed of five immunoglobulin-binding domains, each of which are able to bind proteins from many mammalian species, most notably Immunoglobulin G (IgG) through the heavy chain within the Fc region. While the native form of Protein A was used as the ligand for first generation Protein A resins, the recombinant form (rProtein A) produced in E. coli is the most prevalent today. Modifications to the protein structure of the ligand, the advent of ligands composed of single domain multimers, and multipoint attachment have given rise to the caustic stable, high capacity and extremely robust Protein A resins in use today.

The protein A ligand can either bind directly to the Fc region of an antibody or to an Fc tag that has been fused to the target of interest. Protein A chromatography is a very robust purification procedure and is used as a capture step due to its specificity and, depending on the intended use for the target molecule (antibodies for diagnostic testing), might be the only chromatographic step required to achieve adequate product purity.Toyopearl AF-rProtein A-650F utilizes a new alkaline resistant recombinant Protein A variant. This new non-animal derived ligand is bound to the base bead, Toyopearl HW-65F, using multipoint attachment. This contributes to both the new resin′s alkaline resistance and its low ligand leakages in the range 5-25ng/mg IgG.

Toyopearl AF-rProtein A-650F maintains its capacity well after repeated CIP exposure to 0.1mol/L NaOH and has reasonable tolerance to 0.5mol/L NaOH CIP.

The new resin also tolerates exposure to above ambient temperatures and if needed can be autoclaved.
Good pressure-flow characteristics and kinetics contribute to maintaining good capacity particularly at low residence times.

Specifications

Clean in place with 0.1 M NaOH.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

103.1 °F

Flash Point(C)

39.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service