Skip to Content
Merck
All Photos(1)

Key Documents

819021

TSKgel® Amide-80 (5 µm) HPLC Columns

L x I.D. 1 cm × 4.6 mm, carbamoyl phase

Synonym(s):

TSKgel® Amide-80 Guard Column, 1 cm x 4.6 mm ID, 5 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501
L × I.D.:
1 cm × 4.6 mm
surface area:
450 m2/g
particle size:
5 μm
matrix active group:
carbamoyl phase
pore size:
100 Å
matrix:
silica particle platform
Pricing and availability is not currently available.

Product Name

TSKgel® Amide-80 Guard Column, phase carbamoyl, L × I.D. 1 cm × 4.6 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L68

product line

TSKgel®

manufacturer/tradename

Tosoh 19021

parameter

0-100% organic solvent
10-80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

surface area

450 m2/g

matrix

silica particle platform

matrix active group

carbamoyl phase

particle size

5 μm

pore size

100 Å

operating pH

2.0-7.5

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

General description

Guard Column for 4.6mmID TSKgel Amide-80 columns, 5μm
TSKgel Guard Columns are stand-alone and do not require separate holders.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service